Assay Details
| Gene Symbol: LOC4333804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776892.2 | XP_015632378.1 | 15 - 16 | 1960 | 94 | |
| XM_015776893.2 | XP_015632379.1 | 15 - 16 | 1961 | 94 | ||
| GenBank mRNA | AK059537.1 | - | 1 - 2 | 78 | 94 | |
More Information
Set Membership: |
|

LOC4333804
Genomic Map