Assay Details
| Gene Symbol: LOC4333804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776892.2 | XP_015632378.1 | 19 - 20 | 2303 | 66 | |
| XM_015776893.2 | XP_015632379.1 | 19 - 20 | 2304 | 66 | ||
| GenBank mRNA | AK059537.1 | - | 5 - 6 | 421 | 66 | |
More Information
Set Membership: |
|

LOC4333804
Genomic Map