Assay Details
| Gene Symbol: LOC4333823 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776646.2 | XP_015632132.1 | 3 - 4 | 1038 | 95 | |
| XM_015776648.2 | XP_015632134.1 | 3 - 4 | 1032 | 95 | ||
| GenBank mRNA | AK072689.1 | - | 3 - 4 | 1039 | 95 | |
| AK072869.1 | - | 4 - 5 | 710 | 95 | ||
| DQ389164.1 | - | 3 - 4 | 1032 | 95 | ||
More Information
Set Membership: |
|

LOC4333823
Genomic Map