Assay Details
| Gene Symbol: LOC4333945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776681.2 | XP_015632167.1 | 5 - 6 | 1111 | 136 | |
| GenBank mRNA | AK242106.1 | - | 5 - 6 | 1040 | 136 | |
More Information
Set Membership: |
|

LOC4333945
Genomic Map