Assay Details
| Gene Symbol: LOC4333958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773332.2 | XP_015628818.1 | 2 - 3 | 214 | 70 | |
| XM_015773333.2 | XP_015628819.1 | 1 - 2 | 299 | 70 | ||
| XM_015773334.2 | XP_015628820.1 | 1 - 2 | 298 | 70 | ||
| GenBank mRNA | AK064412.1 | - | 1 - 2 | 96 | 70 | |
| AK106475.1 | - | 1 - 2 | 100 | 70 | ||
| AK108518.1 | - | 1 - 2 | 248 | 70 | ||
| BT131003.1 | - | 1 - 2 | 95 | 70 | ||
More Information
Set Membership: |
|

LOC4333958
Genomic Map