Assay Details
| Gene Symbol: LOC4333958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773332.2 | XP_015628818.1 | 5 - 6 | 443 | 86 | |
| XM_015773333.2 | XP_015628819.1 | 4 - 5 | 528 | 86 | ||
| XM_015773334.2 | XP_015628820.1 | 4 - 5 | 527 | 86 | ||
| GenBank mRNA | AK064412.1 | - | 4 - 5 | 325 | 86 | |
| AK106475.1 | - | 4 - 5 | 329 | 86 | ||
| AK108518.1 | - | 4 - 5 | 477 | 86 | ||
More Information
Set Membership: |
|

LOC4333958
Genomic Map