Assay Details
| Gene Symbol: LOC4333958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773333.2 | XP_015628819.1 | 5 - 6 | 593 | 102 | |
| XM_015773334.2 | XP_015628820.1 | 5 - 6 | 592 | 102 | ||
| GenBank mRNA | AK064412.1 | - | 5 - 6 | 390 | 102 | |
| AK108518.1 | - | 5 - 6 | 542 | 102 | ||
| BT131003.1 | - | 5 - 6 | 298 | 102 | ||
More Information
Set Membership: |
|

LOC4333958
Genomic Map