Assay Details
| Gene Symbol: LOC4333958 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773334.2 | XP_015628820.1 | 8 - 9 | 813 | 58 | |
| GenBank mRNA | AK064412.1 | - | 8 - 9 | 611 | 58 | |
| BT131003.1 | - | 8 - 9 | 519 | 58 | ||
| BT132039.1 | - | 4 - 5 | 237 | 58 | ||
More Information
Set Membership: |
|

LOC4333958
Genomic Map