Assay Details
| Gene Symbol: LOC9269293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795512.2 | XP_015650998.1 | 2 - 3 | 365 | 70 | |
| GenBank mRNA | AK072608.1 | - | 2 - 3 | 310 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015795512.2 transcripts.
Set Membership: |
|

LOC9269293
Genomic Map