Assay Details
| Gene Symbol: LOC9269293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795512.2 | XP_015650998.1 | 3 - 4 | 808 | 61 | |
| GenBank mRNA | AK072608.1 | - | 3 - 4 | 753 | 61 | |
| AK121257.1 | - | 2 - 3 | 1028 | 61 | ||
More Information
Set Membership: |
|

LOC9269293
Genomic Map