Assay Details
| Gene Symbol: LOC9269293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795512.2 | XP_015650998.1 | 6 - 7 | 1758 | 70 | |
| GenBank mRNA | AK072608.1 | - | 6 - 7 | 1699 | 70 | |
| AK121257.1 | - | 5 - 6 | 1978 | 70 | ||
More Information
Set Membership: |
|

LOC9269293
Genomic Map