Assay Details
| Gene Symbol: LOC9268439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015765141.2 | XP_015620627.1 | 4 - 5 | 1169 | 135 | |
| GenBank mRNA | AK067502.1 | - | 4 - 5 | 1171 | 135 | |
More Information
Set Membership: |
|

LOC9268439
Genomic Map