Assay Details
| Gene Symbol: LOC4352218 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015764854.2 | XP_015620340.1 | 3 - 4 | 840 | 62 | |
| GenBank mRNA | AK073151.1 | - | 3 - 4 | 828 | 62 | |
| AK099385.1 | - | 3 - 4 | 831 | 62 | ||
More Information
Set Membership: |
|

LOC4352218
Genomic Map