Assay Details
| Gene Symbol: LOC4330071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK062167.1 | - | 1 | 949 | 149 | |
| AK065162.1 | - | 1 | 984 | 149 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4330071| Gene Symbol: LOC4330071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK062167.1 | - | 1 | 949 | 149 | |
| AK065162.1 | - | 1 | 984 | 149 | ||
Set Membership: |
|