Assay Details
| Gene Symbol: LOC4330104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768303.2 | XP_015623789.1 | 1 - 2 | 462 | 83 | |
| GenBank mRNA | AK101968.1 | - | 1 - 2 | 463 | 83 | |
More Information
Set Membership: |
|

LOC4330104
Genomic Map