Assay Details
| Gene Symbol: LOC4330185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770531.2 | XP_015626017.1 | 1 - 2 | 313 | 70 | |
| GenBank mRNA | AK101944.1 | - | 1 - 2 | 70 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770531.2 transcripts.
Set Membership: |
|

LOC4330185
Genomic Map