Assay Details
| Gene Symbol: LOC4330318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767363.1 | XP_015622849.1 | 10 - 11 | 1108 | 116 | |
| GenBank mRNA | AK060498.1 | - | 7 - 8 | 647 | 116 | |
| AK102210.1 | - | 10 - 11 | 1012 | 116 | ||
More Information
Set Membership: |
|

LOC4330318
Genomic Map