Assay Details
| Gene Symbol: LOC4330318 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767363.1 | XP_015622849.1 | 11 - 12 | 1216 | 75 | |
| GenBank mRNA | AK060498.1 | - | 8 - 9 | 755 | 75 | |
| AK102210.1 | - | 11 - 12 | 1120 | 75 | ||
More Information
Set Membership: |
|

LOC4330318
Genomic Map