Assay Details
| Gene Symbol: LOC4330457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769651.2 | XP_015625137.1 | 5 - 6 | 486 | 95 | |
| GenBank mRNA | AF093629.1 | - | 5 - 6 | 403 | 95 | |
| AK061019.1 | - | 5 - 6 | 464 | 95 | ||
| AK070562.1 | - | 5 - 6 | 439 | 95 | ||
| AK103984.1 | - | 5 - 6 | 431 | 95 | ||
| BT132121.1 | - | 5 - 6 | 459 | 95 | ||
| KF737324.1 | - | 5 - 6 | 404 | 95 | ||
More Information
Set Membership: |
|

LOC4330457
Genomic Map