Assay Details
| Gene Symbol: LOC4330457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769651.2 | XP_015625137.1 | 6 - 7 | 551 | 80 | |
| GenBank mRNA | AK061019.1 | - | 6 - 7 | 529 | 80 | |
| AK070562.1 | - | 6 - 7 | 504 | 80 | ||
| AK103984.1 | - | 6 - 7 | 496 | 80 | ||
| BT132121.1 | - | 6 - 7 | 524 | 80 | ||
| KF737324.1 | - | 6 - 7 | 469 | 80 | ||
More Information
Set Membership: |
|

LOC4330457
Genomic Map