Assay Details
| Gene Symbol: LOC4330830 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768164.1 | XP_015623650.1 | 4 | 1429 | 124 | |
| GenBank mRNA | EU267975.1 | - | 4 | 1410 | 124 | |
More Information
Set Membership: |
|

LOC4330830
Genomic Map