Assay Details
| Gene Symbol: LOC4331062 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771759.2 | XP_015627245.1 | 1 - 2 | 437 | 65 | |
| GenBank mRNA | AK071591.1 | - | 1 - 2 | 413 | 65 | |
| AK243048.1 | - | 1 - 2 | 413 | 65 | ||
More Information
Set Membership: |
|

LOC4331062
Genomic Map