Assay Details
| Gene Symbol: LOC4331389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776725.2 | XP_015632211.1 | 7 - 8 | 1713 | 69 | |
| GenBank mRNA | AB807477.1 | - | 7 - 8 | 1326 | 69 | |
| AK060853.1 | - | 4 - 5 | 854 | 69 | ||
| AK065394.1 | - | 7 - 8 | 1528 | 69 | ||
More Information
Set Membership: |
|

LOC4331389
Genomic Map