Assay Details
| Gene Symbol: LOC4332016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777402.2 | XP_015632888.1 | 6 - 7 | 618 | 109 | |
| GenBank mRNA | AK059829.1 | - | 6 - 7 | 573 | 109 | |
More Information
Set Membership: |
|

LOC4332016
Genomic Map