Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 40 - 41 | 5426 | 135 | |
| GenBank mRNA | AK099757.1 | - | 19 - 20 | 2321 | 135 | |
| AK120639.1 | - | 16 - 17 | 1851 | 135 | ||
More Information
Set Membership: |
|

LOC4332045
Genomic Map