Assay Details
| Gene Symbol: LOC4332045 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774461.2 | XP_015629947.1 | 22 - 23 | 3170 | 83 | |
| GenBank mRNA | AK099757.1 | - | 1 - 2 | 65 | 83 | |
More Information
Set Membership: |
|

LOC4332045
Genomic Map