Assay Details
| Gene Symbol: LOC4332108 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776193.1 | XP_015631679.1 | 5 - 6 | 1031 | 85 | |
| GenBank mRNA | AB180688.1 | - | 5 - 6 | 1031 | 85 | |
| AK243037.1 | - | 5 - 6 | 451 | 85 | ||
| X14244.1 | - | 5 - 6 | 433 | 85 | ||
More Information
Set Membership: |
|

LOC4332108
Genomic Map