Assay Details
| Gene Symbol: LOC4332121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773277.2 | XP_015628763.1 | 1 | 1840 | 135 | |
| GenBank mRNA | AK061758.1 | - | 1 | 1724 | 135 | |
| AK066138.1 | - | 1 | 1847 | 135 | ||
More Information
Set Membership: |
|

LOC4332121
Genomic Map