Assay Details
| Gene Symbol: LOC4332477 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775503.2 | XP_015630989.1 | 3 - 4 | 565 | 60 | |
| GenBank mRNA | AK066644.1 | - | 3 - 4 | 571 | 60 | |
More Information
Set Membership: |
|

LOC4332477
Genomic Map