Assay Details
| Gene Symbol: LOC4332482 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773501.2 | XP_015628987.1 | 7 - 8 | 693 | 74 | |
| GenBank mRNA | AK100125.1 | - | 7 - 8 | 680 | 74 | |
| KY170874.1 | - | 7 - 8 | 545 | 74 | ||
More Information
Set Membership: |
|

LOC4332482
Genomic Map