Assay Details
| Gene Symbol: LOC4332918 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776154.2 | XP_015631640.1 | 12 - 13 | 1337 | 75 | |
| GenBank mRNA | AK065282.1 | - | 12 - 13 | 1259 | 75 | |
| AK122008.1 | - | 12 - 13 | 1359 | 75 | ||
More Information
Set Membership: |
|

LOC4332918
Genomic Map