Assay Details
| Gene Symbol: LOC4333571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776175.2 | XP_015631661.1 | 22 - 23 | 3012 | 104 | |
| GenBank mRNA | AK111443.1 | - | 5 - 6 | 940 | 104 | |
More Information
Set Membership: |
|

LOC4333571
Genomic Map