Assay Details
Gene Symbol: LOC4333612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776119.1 | XP_015631605.1 | 2 - 3 | 3127 | 64 | |
GenBank mRNA | AK073458.1 | - | 2 - 3 | 518 | 64 | |
AK120058.1 | - | 2 - 3 | 2987 | 64 | ||
AK120778.1 | - | 2 - 3 | 3136 | 64 | ||
BT130682.1 | - | 2 - 3 | 493 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
