Assay Details
| Gene Symbol: LOC4333961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775701.1 | XP_015631187.1 | 8 - 9 | 3455 | 69 | |
| GenBank mRNA | AK064260.1 | - | 1 - 2 | 358 | 69 | |
| AK119472.1 | - | 1 - 2 | 1419 | 69 | ||
More Information
Set Membership: |
|

LOC4333961
Genomic Map