Assay Details
| Gene Symbol: LOC4334298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775434.2 | XP_015630920.1 | 6 - 7 | 3633 | 71 | |
| GenBank mRNA | AK105528.1 | - | 3 - 4 | 369 | 71 | |
| AK120460.1 | - | 3 - 4 | 2263 | 71 | ||
More Information
Set Membership: |
|

LOC4334298
Genomic Map