Assay Details
| Gene Symbol: LOC4334349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061286.1 | - | 2 - 3 | 383 | 69 | |
| AK073187.1 | - | 2 - 3 | 379 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4334349| Gene Symbol: LOC4334349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK061286.1 | - | 2 - 3 | 383 | 69 | |
| AK073187.1 | - | 2 - 3 | 379 | 69 | ||
Set Membership: |
|