Assay Details
Gene Symbol: LOC4334394 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015777656.2 | XP_015633142.1 | 2 | 772 | 88 | |
XM_015777657.2 | XP_015633143.1 | 1 | 3243 | 88 | ||
GenBank mRNA | AK108205.1 | - | 1 | 51 | 88 | |
AY526867.1 | - | 1 | 51 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015777657.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
