Assay Details
| Gene Symbol: LOC4334469 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776480.1 | XP_015631966.1 | 6 - 7 | 1041 | 149 | |
| GenBank mRNA | AK288909.1 | - | 6 - 7 | 1041 | 149 | |
More Information
Set Membership: |
|

LOC4334469
Genomic Map