Assay Details
| Gene Symbol: LOC4334751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774199.2 | XP_015629685.1 | 11 - 12 | 2504 | 107 | |
| XM_015774200.2 | XP_015629686.1 | 11 - 12 | 2530 | 107 | ||
| XM_015774201.2 | XP_015629687.1 | 12 - 13 | 2121 | 107 | ||
| GenBank mRNA | AK068216.1 | - | 11 - 12 | 2511 | 107 | |
| AK101493.1 | - | 11 - 12 | 2511 | 107 | ||
More Information
Set Membership: |
|

LOC4334751
Genomic Map