Assay Details
| Gene Symbol: LOC4334751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774199.2 | XP_015629685.1 | 9 - 10 | 2150 | 110 | |
| XM_015774200.2 | XP_015629686.1 | 9 - 10 | 2176 | 110 | ||
| XM_015774201.2 | XP_015629687.1 | 10 - 11 | 1767 | 110 | ||
| GenBank mRNA | AK068216.1 | - | 9 - 10 | 2157 | 110 | |
| AK101493.1 | - | 9 - 10 | 2157 | 110 | ||
More Information
Set Membership: |
|

LOC4334751
Genomic Map