Assay Details
| Gene Symbol: LOC4334845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775841.1 | XP_015631327.1 | 2 - 3 | 788 | 66 | |
| GenBank mRNA | AK062137.1 | - | 2 - 3 | 603 | 66 | |
| AK098951.1 | - | 2 - 3 | 606 | 66 | ||
More Information
Set Membership: |
|

LOC4334845
Genomic Map