Assay Details
| Gene Symbol: LOC4334845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775841.1 | XP_015631327.1 | 7 - 8 | 1186 | 69 | |
| GenBank mRNA | AK062137.1 | - | 7 - 8 | 1001 | 69 | |
| AK098951.1 | - | 7 - 8 | 1004 | 69 | ||
More Information
Set Membership: |
|

LOC4334845
Genomic Map