Assay Details
| Gene Symbol: LOC4334873 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775319.2 | XP_015630805.1 | 1 | 247 | 54 | |
| GenBank mRNA | AK109024.1 | - | 1 | 253 | 54 | |
More Information
Set Membership: |
|

LOC4334873
Genomic Map