Assay Details
| Gene Symbol: LOC4334912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780670.2 | XP_015636156.1 | 4 - 5 | 840 | 108 | |
| GenBank mRNA | AK059655.1 | - | 4 - 5 | 770 | 108 | |
| AK101488.1 | - | 4 - 5 | 773 | 108 | ||
More Information
Set Membership: |
|

LOC4334912
Genomic Map