Assay Details
| Gene Symbol: LOC4334912 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015780670.2 | XP_015636156.1 | 5 - 6 | 961 | 95 | |
| GenBank mRNA | AK059655.1 | - | 5 - 6 | 891 | 95 | |
| AK101488.1 | - | 5 - 6 | 894 | 95 | ||
More Information
Set Membership: |
|

LOC4334912
Genomic Map