Assay Details
| Gene Symbol: LOC4335090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781606.2 | XP_015637092.1 | 1 - 2 | 250 | 75 | |
| XM_015781607.2 | XP_015637093.1 | 1 - 2 | 252 | 75 | ||
| GenBank mRNA | AB066270.1 | - | 1 - 2 | 250 | 75 | |
More Information
Set Membership: |
|

LOC4335090
Genomic Map