Assay Details
| Gene Symbol: LOC4335090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781606.2 | XP_015637092.1 | 6 - 7 | 1153 | 98 | |
| XM_015781607.2 | XP_015637093.1 | 6 - 7 | 1155 | 98 | ||
| GenBank mRNA | AB066270.1 | - | 6 - 7 | 1153 | 98 | |
| AK100631.1 | - | 5 - 6 | 1350 | 98 | ||
| AK121319.1 | - | 2 - 3 | 355 | 98 | ||
More Information
Set Membership: |
|

LOC4335090
Genomic Map