Assay Details
| Gene Symbol: LOC4335090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781606.2 | XP_015637092.1 | 9 - 10 | 1617 | 61 | |
| XM_015781607.2 | XP_015637093.1 | 9 - 10 | 1619 | 61 | ||
| GenBank mRNA | AK100631.1 | - | 8 - 9 | 1814 | 61 | |
| AK121319.1 | - | 5 - 6 | 819 | 61 | ||
More Information
Set Membership: |
|

LOC4335090
Genomic Map