Assay Details
| Gene Symbol: LOC4335427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779634.2 | XP_015635120.1 | 6 - 7 | 733 | 101 | |
| GenBank mRNA | AK102927.1 | - | 6 - 7 | 735 | 101 | |
| AY177695.2 | - | 6 - 7 | 569 | 101 | ||
More Information
Set Membership: |
|

LOC4335427
Genomic Map