Assay Details
| Gene Symbol: LOC4335427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015779634.2 | XP_015635120.1 | 7 - 8 | 111 | ||
| GenBank mRNA | AK102927.1 | - | 7 - 8 | 111 | ||
| AY177695.2 | - | 7 - 8 | 725 | 110 | ||
More Information
Set Membership: |
|

LOC4335427
Genomic Map